1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)piperazin
Lieferant: BIONET RESEARCH
8F-928.500
8F-928.1
8F-928.5
8F-928.10
BIOT8F-928.500EA
0
EUR
BIOT8F-928.500
BIOT8F-928.1
BIOT8F-928.5
BIOT8F-928.10
1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)piperazin
1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)piperazin
Formel:
C₁₀H₁₁ClF₃N₃ |
MDL:
MFCD00173934 CAS-Nummer: 132834-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...