1-[3-(Trifluoromethyl)pyridin-2-yl]piperazine

Lieferant: FLUOROCHEM

Synonyme: 2-(Piperazin-1-yl)-3-(trifluoromethyl)pyridine, 1-(3-(Trifluoromethyl)pyridin-2-yl)piperazine

008457-1G 008457-5G
FLUO008457-1GEA 73.1 EUR
FLUO008457-1G FLUO008457-5G
1-[3-(Trifluoromethyl)pyridin-2-yl]piperazine
1-[3-(Trifluoromethyl)pyridin-2-yl]piperazine
Formel: C₁₀H₁₂F₃N₃
Molecular Weight: 231,218 g/mol
MDL: MFCD00114705
CAS-Nummer: 87394-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR