5-(4-Pyridyl)-1H-tetrazol

Lieferant: FLUOROCHEM

Synonyme: 4-(1H-Tetrazol-5-yl)pyridine

CFF680.1 005513-1G 005513-5G
CHMPCFF680.1EA 532 EUR
CHMPCFF680.1 FLUO005513-1G FLUO005513-5G
5-(4-Pyridyl)-1H-tetrazol
5-(4-Pyridyl)-1H-tetrazole
Formel: C₆H₅N₅
Molecular Weight: 147.14 g/mol
MDL: MFCD00068115
CAS-Nummer: 14389-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR