6-Bromtetrazolo[1,5-a]pyridin

Lieferant: COMBI-BLOCKS

HI-1306-5G HI-1306-1G
COBBHI-1306-5GEA 0 EUR
COBBHI-1306-5G COBBHI-1306-1G
6-Bromtetrazolo[1,5-a]pyridin
6-Bromtetrazolo[1,5-a]pyridin
Formel: C₅H₃BrN₄
MDL: MFCD09258642
CAS-Nummer: 35235-74-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR