(1-Phenyl-1H-1,2,3-triazol-4-yl)methanol

Lieferant: BIONET RESEARCH

4F-411S.500 4F-411S.1 4F-411S.5 4F-411S.10
BIOT4F-411S.500EA 0 EUR
BIOT4F-411S.500 BIOT4F-411S.1 BIOT4F-411S.5 BIOT4F-411S.10
(1-Phenyl-1H-1,2,3-triazol-4-yl)methanol
(1-Phenyl-1H-1,2,3-triazol-4-yl)methanol
Formel: C₉H₉N₃O
Molecular Weight: 175,1900 g/mol
MDL: MFCD00100214
CAS-Nummer: 103755-58-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR