4-(4-Bromphenoxy)piperidin

Lieferant: CHEMPUR

60-0133.500 60-0133.1
CHMP60-0133.500EA 0 EUR
CHMP60-0133.500 CHMP60-0133.1
4-(4-Bromphenoxy)piperidin
4-(4-Bromphenoxy)piperidin
Formel: C₁₁H₁₄BrNO
MDL: MFCD04115016
CAS-Nummer: 74130-05-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR