4-(3-Bromphenoxy)piperidin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-2095-5G HC-2095-1G
COBBHC-2095-5GEA 1490 EUR
COBBHC-2095-5G COBBHC-2095-1G
4-(3-Bromphenoxy)piperidin Hydrochlorid
4-(3-Bromphenoxy)piperidin Hydrochlorid
Formel: C₁₁H₁₅BrClNO
Molecular Weight: 292.599 g/mol
MDL: MFCD09879219
CAS-Nummer: 1072944-50-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR