4-(4-Chlorphenoxy)piperidin

Lieferant: CHEMPUR

60-0051.500 60-0051.1
CHMP60-0051.500EA 0 EUR
CHMP60-0051.500 CHMP60-0051.1
4-(4-Chlorphenoxy)piperidin
4-(4-Chlorphenoxy)piperidin
Formel: C₁₁H₁₄ClNO
Molecular Weight: 211,69 g/mol
MDL: MFCD04113544
CAS-Nummer: 97839-99-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR