1-Boc-4-(4-nitrophenoxy)piperidin

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 4-(4-nitrophenoxy)piperidine-1-carboxylate

AN-1499-5G AN-1499-1G
COBBAN-1499-5GEA 519 EUR
COBBAN-1499-5G COBBAN-1499-1G
1-Boc-4-(4-nitrophenoxy)piperidin
1-Boc-4-(4-nitrophenoxy)piperidin
Formel: C₁₆H₂₂N₂O₅
Molecular Weight: 322.3563 g/mol
MDL: MFCD06200882
CAS-Nummer: 138227-62-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR