4-(4-Fluorphenoxy)piperidin

Lieferant: CHEMPUR

60-0132.500 60-0132.1
CHMP60-0132.500EA 0 EUR
CHMP60-0132.500 CHMP60-0132.1
4-(4-Fluorphenoxy)piperidin
4-(4-Fluorphenoxy)piperidin
Formel: C₁₁H₁₄FNO
Molecular Weight: 195,2334 g/mol
MDL: MFCD04115015
CAS-Nummer: 3413-28-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR