4-(4-Nitrophenoxy)piperidin

Lieferant: CHEMPUR

60-0140.500 60-0140.1
CHMP60-0140.500EA 0 EUR
CHMP60-0140.500 CHMP60-0140.1
4-(4-Nitrophenoxy)piperidin
4-(4-Nitrophenoxy)piperidin
Formel: C₁₁H₁₄N₂O₃
MDL: MFCD04115022
CAS-Nummer: 162402-39-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR