4-(2-Nitrophenoxy)piperidin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-2094-5G HC-2094-1G
COBBHC-2094-5GEA 451 EUR
COBBHC-2094-5G COBBHC-2094-1G
4-(2-Nitrophenoxy)piperidin Hydrochlorid
4-(2-Nitrophenoxy)piperidin Hydrochlorid
Formel: C₁₁H₁₅ClN₂O₃
MDL: MFCD09909466
CAS-Nummer: 1072944-49-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR