4-(4-Methoxyphenoxy)piperidin Hydrochlorid
Lieferant: COMBI-BLOCKS
HC-7004-5G
HC-7004-1G
COBBHC-7004-5GEA
358
EUR
COBBHC-7004-5G
COBBHC-7004-1G
4-(4-Methoxyphenoxy)piperidin Hydrochlorid
4-(4-Methoxyphenoxy)piperidin Hydrochlorid
Formel:
C₁₂H₁₈ClNO₂ |
MDL:
MFCD03840124 CAS-Nummer: 333954-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...