3-Phenoxypiperidin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H50912.03 H50912.MD
H50912.03EA 287 EUR
H50912.03 H50912.MD
3-Phenoxypiperidin ≥97%
3-Phenoxypiperidin

Purity: 97% CAS No.: 151666-08-9 Molecular Formula: C11H15NO Formula Weight: 177.25 Refractive Index: 1.5430

Formel: C₁₁H₁₅NO
Molecular Weight: 177.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04117746
CAS-Nummer: 151666-08-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR