4-Phenoxypiperidin

Lieferant: CHEMPUR

60-0061.1 60-0061.5
CHMP60-0061.1EA 0 EUR
CHMP60-0061.1 CHMP60-0061.5
4-Phenoxypiperidin
4-Phenoxypiperidin
Formel: C₁₁H₁₅NO
Molecular Weight: 177.25 g/mol
MDL: MFCD04114971
CAS-Nummer: 3202-33-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR