2-Brominden ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B25474.14
B25474.06
B25474.14EA
146
EUR
B25474.14
B25474.06
2-Brominden ≥98%
2-Brominden
Purity: 98%
CAS No.: 10485-09-3
Molecular Formula: C9H7Br
Formula Weight: 195.06
Melting Point: 35-37°
MDL No.: MFCD06797863
Formel:
C₉H₇Br Molecular Weight: 195.06 g/mol Schmelzpunkt: 35…37 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797863 CAS-Nummer: 10485-09-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...