1-(4-Cyanphenyl)piperazin

Lieferant: BIONET RESEARCH

4R-0247.500 4R-0247.1 4R-0247.5 4R-0247.10
BIOT4R-0247.500EA 0 EUR
BIOT4R-0247.500 BIOT4R-0247.1 BIOT4R-0247.5 BIOT4R-0247.10
1-(4-Cyanphenyl)piperazin
1-(4-Cyanphenyl)piperazin
Formel: C₁₁H₁₃N₃
MDL: MFCD01320890
CAS-Nummer: 68104-63-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR