1-(4-Cyanphenyl)piperazin
Lieferant: FLUOROCHEM
Synonyme:
4-(Piperazin-1-yl)benzonitrile, 4-(1-Piperazinyl)benzonitrile, 1-(4-Cyanophenyl)piperazine
018123-1G
018123-5G
FLUO018123-1GEA
51.5
EUR
FLUO018123-1G
FLUO018123-5G
1-(4-Cyanphenyl)piperazin
1-(4-Cyanphenyl)piperazin
Formel:
C₁₁H₁₃N₃ Molecular Weight: 187,2410 g/mol |
MDL:
MFCD01320890 CAS-Nummer: 68104-63-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...