2-(tert-Butyldimethylsilyloxy)-6-bromnaphthalin

Lieferant: COMBI-BLOCKS

OT-0474-5G OT-0474-1G OT-0474-25G
COBBOT-0474-5GEA 74.2 EUR
COBBOT-0474-5G COBBOT-0474-1G COBBOT-0474-25G
2-(tert-Butyldimethylsilyloxy)-6-bromnaphthalin
2-(tert-Butyldimethylsilyloxy)-6-bromnaphthalin
Formel: C₁₆H₂₁BrOSi
MDL: MFCD11044858
CAS-Nummer: 100751-65-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR