5-Brom-(2,3-methylendioxy)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(6-Bromobenzo[d][1,3]dioxol-4-yl)boronic acid
BB-5462-5G
BB-5462-1G
BB-5462-250M
COBBBB-5462-5GEA
2070
EUR
COBBBB-5462-5G
COBBBB-5462-1G
COBBBB-5462-250M
5-Brom-(2,3-methylendioxy)phenylboronsäure
5-Brom-(2,3-methylendioxy)phenylboronsäure
Formel:
C₇H₆BBrO₄ Molecular Weight: 244,8400 g/mol |
MDL:
MFCD11855838 CAS-Nummer: 1150114-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...