Bis(tributylstannyl)acetylen

Lieferant: CHEMPUR
Gefahr

FL11018.POR 011018-1G 011018-5G
CHMPFL11018.POREA 0 EUR
CHMPFL11018.POR FLUO011018-1G FLUO011018-5G
Bis(tributylstannyl)acetylen
Bis(tributylstannyl)acetylen
Formel: C₂₆H₅₄Sn₂
MDL: MFCD00010239
CAS-Nummer: 994-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR