1-(5-Chlor-2-pyridinyl)piperazin ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-(5-Chloro-2-pyridyl)piperazine

H50862.03 H50862.MD
H50862.03EA 179 EUR
H50862.03 H50862.MD
1-(5-Chlor-2-pyridinyl)piperazin ≥99%
1-(5-Chlor-2-pyridinyl)piperazin

Purity: 99% CAS No.: 87394-65-8 Molecular Formula: C9H12ClN3 Formula Weight: 197.67 MDL No.: MFCD04115049

Formel: C₉H₁₂ClN₃
Molecular Weight: 197,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115049
CAS-Nummer: 87394-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR