1-(3,5-Dichlor-4-pyridinyl)piperazin

Lieferant: FLUOROCHEM

017238-2.5G 017238-1G
FLUO017238-2.5GEA 0 EUR
FLUO017238-2.5G FLUO017238-1G
1-(3,5-Dichlor-4-pyridinyl)piperazin
1-(3,5-Dichlor-4-pyridinyl)piperazin
Formel: C₉H₁₁Cl₂N₃
Molecular Weight: 232.11 g/mol
MDL: MFCD00221400
CAS-Nummer: 175277-80-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR