1,2,4-Triazol-3-carbonsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1H-1,2,4-Triazole-3-carboxylic acid
M-Clarity™ Qualitätsstufe = MQ100
658472-10G
658472-1G
SIAL658472-10GEA
139
EUR
SIAL658472-10G
SIAL658472-1G
1,2,4-Triazol-3-carbonsäure, Sigma-Aldrich®
1,2,4-Triazole-3-carboxylic acid
Formel:
C₃H₃N₃O₂ Molecular Weight: 113.0748 g/mol |
MDL:
MFCD00078268 CAS-Nummer: 4928-87-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...