3-(Propylthio)-1H-1,2,4-triazol

Lieferant: CHEMPUR

FL9882.POR 009882-1G 009882-5G
CHMPFL9882.POREA 0 EUR
CHMPFL9882.POR FLUO009882-1G FLUO009882-5G
3-(Propylthio)-1H-1,2,4-triazol
3-(Propylthio)-1H-1,2,4-triazol
Formel: C₅H₉N₃S
CAS-Nummer: 34945-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR