3-(Propylthio)-1H-1,2,4-triazol
Lieferant: Thermo Scientific
JFD03131.1
JFD03131.10
MAYBJFD03131.1EA
0
EUR
MAYBJFD03131.1
MAYBJFD03131.10
3-(Propylthio)-1H-1,2,4-triazol
3-(Propylthio)-1H-1,2,4-triazol
Formel:
C₅H₉N₃S |
CAS-Nummer:
34945-15-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...