1-Acetamido-4-bromnaphthalin

Lieferant: COMBI-BLOCKS

OT-0413-1G OT-0413-5G
COBBOT-0413-1GEA 127 EUR
COBBOT-0413-1G COBBOT-0413-5G
1-Acetamido-4-bromnaphthalin
1-Acetamido-4-bromnaphthalin
Formel: C₁₂H₁₀BrNO
MDL: MFCD00085962
CAS-Nummer: 91394-66-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR