(E)-1-Hepten-1,2-diboronsäurebis(pinakol)ester
Lieferant: FLUOROCHEM
010763-2G
010763-5G
FLUO010763-2GEA
0
EUR
FLUO010763-2G
FLUO010763-5G
(E)-1-Hepten-1,2-diboronsäurebis(pinakol)ester
(E)-1-Hepten-1,2-diboronsäurebis(pinakol)ester
Formel:
C₁₉H₃₆B₂O₄ |
MDL:
MFCD02683463 CAS-Nummer: 307531-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...