(E)-1-Penten-1,2-diboronsäurebis(pinakol)ester

Lieferant: FLUOROCHEM

010765-2G 010765-5G
FLUO010765-2GEA 0 EUR
FLUO010765-2G FLUO010765-5G
(E)-1-Penten-1,2-diboronsäurebis(pinakol)ester
(E)-1-Penten-1,2-diboronsäurebis(pinakol)ester
Formel: C₁₇H₃₂B₂O₄
MDL: MFCD03093909
CAS-Nummer: 307531-75-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR