trans-3,3-Dimethyl-1-butenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 2-tert-Butyl-trans-vinylboronic acid, (E)-(3,3-Dimethylbut-1-en-1-yl)boronic acid

016252-1G 016252-5G
FLUO016252-1GEA 377 EUR
FLUO016252-1G FLUO016252-5G
trans-3,3-Dimethyl-1-butenylboronsäure
trans-3,3-Dimethyl-1-butenylboronic acid
Formel: C₆H₁₃BO₂
Molecular Weight: 127,98 g/mol
MDL: MFCD01074624
CAS-Nummer: 86595-37-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR