2-Cyan-5-nitrophenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
4-Nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
PN-5756-1G
PN-5756-250M
COBBPN-5756-1GEA
595
EUR
COBBPN-5756-1G
COBBPN-5756-250M
2-Cyan-5-nitrophenylboronsäurepinakolester
2-Cyan-5-nitrophenylboronsäurepinakolester
Formel:
C₁₃H₁₅BN₂O₄ Molecular Weight: 274.0802 g/mol |
MDL:
MFCD12546472 CAS-Nummer: 1218791-28-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...