4-(N,N-Dimethylamino)-3-nitrophenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
N,N-Dimethyl-2-nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
PN-5698-1G
PN-5698-250M
COBBPN-5698-1GEA
312
EUR
COBBPN-5698-1G
COBBPN-5698-250M
4-(N,N-Dimethylamino)-3-nitrophenylboronsäurepinakolester
4-(N,N-Dimethylamino)-3-nitrophenylboronsäurepinakolester
Formel:
C₁₄H₂₁BN₂O₄ Molecular Weight: 292.1400 g/mol |
MDL:
MFCD12546469 CAS-Nummer: 1218791-24-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...