3-Cyan-5-nitrophenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Cyano-5-nitrophenyl)boronic acid
BB-2829-5G
BB-2829-1G
COBBBB-2829-5GEA
1750
EUR
COBBBB-2829-5G
COBBBB-2829-1G
3-Cyan-5-nitrophenylboronsäure
3-Cyan-5-nitrophenylboronsäure
Formel:
C₇H₅BN₂O₄ Molecular Weight: 191,936 g/mol |
MDL:
MFCD03411554 CAS-Nummer: 913835-33-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...