4-Cyan-2-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Cyano-2-nitrophenyl)boronic acid

BB-2046-1G BB-2046-5G
COBBBB-2046-1GEA 182 EUR
COBBBB-2046-1G COBBBB-2046-5G
4-Cyan-2-nitrophenylboronsäure
4-Cyan-2-nitrophenylboronsäure
Formel: C₇H₅BN₂O₄
Molecular Weight: 191,9400 g/mol
MDL: MFCD04115646
CAS-Nummer: 850568-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR