3-Acetamido-2-nitrophenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Acetamido-2-nitrobenzeneboronic acid
BB-2217-1G
BB-2217-250M
COBBBB-2217-1GEA
1360
EUR
COBBBB-2217-1G
COBBBB-2217-250M
3-Acetamido-2-nitrophenylboronsäure
3-Acetamido-2-nitrophenylboronsäure
Formel:
C₈H₉BN₂O₅ Molecular Weight: 223,98 g/mol |
MDL:
MFCD06659828 CAS-Nummer: 78887-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...