3-(Cyclopropylcarbamoyl)-5-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-(Cyclopropylcarbamoyl)-5-nitrophenyl)boronic acid

BB-2834-5G BB-2834-1G
COBBBB-2834-5GEA 581 EUR
COBBBB-2834-5G COBBBB-2834-1G
3-(Cyclopropylcarbamoyl)-5-nitrophenylboronsäure
3-(Cyclopropylcarbamoyl)-5-nitrophenylboronsäure
Formel: C₁₀H₁₁BN₂O₅
Molecular Weight: 250,0200 g/mol
MDL: MFCD07783878
CAS-Nummer: 871332-86-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR