3-Amino-5-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Amino-5-nitrobenzeneboronic acid

BB-3412-5G BB-3412-1G
COBBBB-3412-5GEA 2070 EUR
COBBBB-3412-5G COBBBB-3412-1G
3-Amino-5-nitrophenylboronsäure
3-Amino-5-nitrophenylboronsäure
Formel: C₆H₇BN₂O₄
Molecular Weight: 181.94 g/mol
MDL: MFCD08436012
CAS-Nummer: 89466-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR