3-(Cyclopentyloxy)-4-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-(Cyclopentyloxy)-4-methoxyphenyl)boronic acid

BB-9036-1G BB-9036-250M
COBBBB-9036-1GEA 623 EUR
COBBBB-9036-1G COBBBB-9036-250M
3-(Cyclopentyloxy)-4-methoxyphenylboronsäure
3-(Cyclopentyloxy)-4-methoxyphenylboronsäure
Formel: C₁₂H₁₇BO₄
Molecular Weight: 236,07 g/mol
MDL: MFCD04039013
CAS-Nummer: 159613-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR