2,6-Di(dimethylamino)benzonitril

Lieferant: Thermo Scientific

CD08880.1 CD08880.10
MAYBCD08880.1EA 0 EUR
MAYBCD08880.1 MAYBCD08880.10
2,6-Di(dimethylamino)benzonitril
2,6-Di(dimethylamino)benzonitril
Formel: C₁₁H₁₅N₃
CAS-Nummer: 20926-04-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR