3-(Cyclopentyloxy)-5-(trifluormethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-6377-1G BB-6377-5G
COBBBB-6377-1GEA 303 EUR
COBBBB-6377-1G COBBBB-6377-5G
3-(Cyclopentyloxy)-5-(trifluormethoxy)phenylboronsäure
3-(Cyclopentyloxy)-5-(trifluormethoxy)phenylboronsäure
Formel: C₁₂H₁₄BF₃O₄
MDL: MFCD16295065
CAS-Nummer: 1256346-04-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR