3-Hydroxy-5-(trifluormethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3974-5G BB-3974-1G
COBBBB-3974-5GEA 0 EUR
COBBBB-3974-5G COBBBB-3974-1G
3-Hydroxy-5-(trifluormethoxy)phenylboronsäure
3-Hydroxy-5-(trifluormethoxy)phenylboronsäure
Formel: C₇H₆BF₃O₄
MDL: MFCD09475898
CAS-Nummer: 957120-22-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR