2-(Methylthio)-5-(trifluormethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5047-5G BB-5047-1G
COBBBB-5047-5GEA 2230 EUR
COBBBB-5047-5G COBBBB-5047-1G
2-(Methylthio)-5-(trifluormethoxy)phenylboronsäure
2-(Methylthio)-5-(trifluormethoxy)phenylboronsäure
Formel: C₈H₈BF₃O₃S
MDL: MFCD09800888
CAS-Nummer: 957121-11-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR