3-Isopropoxy-5-(trifluormethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Isopropoxy-5-(trifluoromethoxy)phenyl)boronic acid

BB-6374-1G BB-6374-250M
COBBBB-6374-1GEA 0 EUR
COBBBB-6374-1G COBBBB-6374-250M
3-Isopropoxy-5-(trifluormethoxy)phenylboronsäure
3-Isopropoxy-5-(trifluormethoxy)phenylboronsäure
Formel: C₁₀H₁₂BF₃O₄
Molecular Weight: 264.0100 g/mol
MDL: MFCD16295063
CAS-Nummer: 1256346-01-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR