3-Ethoxy-5-(trifluormethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-6330-1G BB-6330-5G BB-6330-250M
COBBBB-6330-1GEA 303 EUR
COBBBB-6330-1G COBBBB-6330-5G COBBBB-6330-250M
3-Ethoxy-5-(trifluormethoxy)phenylboronsäure
3-Ethoxy-5-(trifluormethoxy)phenylboronsäure
Formel: C₉H₁₀BF₃O₄
MDL: MFCD16295059
CAS-Nummer: 1256345-87-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR