(4-Fluorphenylthio)acetonitril

Lieferant: FLUOROCHEM

Synonyme: 2-((4-Fluorophenyl)thio)acetonitrile

006344-1G 006344-5G
FLUO006344-1GEA 87.9 EUR
FLUO006344-1G FLUO006344-5G
(4-Fluorphenylthio)acetonitril
(4-Fluorphenylthio)acetonitril
Formel: C₈H₆FNS
Molecular Weight: 167,203 g/mol
MDL: MFCD00179342
CAS-Nummer: 18527-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR