(4-Fluorphenylthio)acetonitril

Lieferant: Thermo Scientific

Synonyme: 2-((4-Fluorophenyl)thio)acetonitrile

KM01391.1 KM01391.10 KM01391.50
MAYBKM01391.1EA 0 EUR
MAYBKM01391.1 MAYBKM01391.10 MAYBKM01391.50
(4-Fluorphenylthio)acetonitril
(4-Fluorphenylthio)acetonitril
Formel: C₈H₆FNS
Molecular Weight: 167,203 g/mol
MDL: MFCD00179342
CAS-Nummer: 18527-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR