3-Acetylphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone, 3-Acetylphenylboronic acid, pinacol ester

PN-2186-5G PN-2186-1G
COBBPN-2186-5GEA 379 EUR
COBBPN-2186-5G COBBPN-2186-1G
3-Acetylphenylboronsäurepinakolester
3-Acetylphenylboronsäurepinakolester
Formel: C₁₄H₁₉BO₃
Molecular Weight: 246,1099 g/mol
MDL: MFCD05863923
CAS-Nummer: 214360-49-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR