4-Acetylphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone, 4-Acetylphenylboronic acid, pinacol ester, 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone

PN-2187-5G PN-2187-1G
COBBPN-2187-5GEA 223 EUR
COBBPN-2187-5G COBBPN-2187-1G
4-Acetylphenylboronsäurepinakolester
4-Acetylphenylboronsäurepinakolester
Formel: C₁₄H₁₉BO₃
Molecular Weight: 246.1099 g/mol
MDL: MFCD05863922
CAS-Nummer: 171364-81-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR