3-(Cyclopentyloxy)-5-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-(Cyclopentyloxy)-5-methylphenyl)boronic acid

BB-6383-5G BB-6383-1G BB-6383-250M
COBBBB-6383-5GEA 1810 EUR
COBBBB-6383-5G COBBBB-6383-1G COBBBB-6383-250M
3-(Cyclopentyloxy)-5-methylphenylboronsäure
3-(Cyclopentyloxy)-5-methylphenylboronsäure
Formel: C₁₂H₁₇BO₃
Molecular Weight: 220,0700 g/mol
MDL: MFCD16295068
CAS-Nummer: 1256346-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR