4-(Aminomethyl)benzonitril Hydrochlorid 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-(Aminomethyl)benzonitrile HCl, α-Amino-p-tolunitrile Hydrochloride, 4-(Aminomethyl)benzonitrile Hydrochloride, 4-Cyanobenzylamine Hydrochloride
H27473.14
H27473.06
H27473.14EA
288
EUR
H27473.14
H27473.06
4-(Aminomethyl)benzonitril Hydrochlorid 97%
4-(Aminomethyl)benzonitril Hydrochlorid
Purity: 97%
CAS No.: 15996-76-6
Molecular Formula: C8H8N2.HCl
Formula Weight: 168.62
Melting Point: 279-284°
MDL No.: MFCD01861472
Formel:
C₈H₉ClN₂ Molecular Weight: 168.62 g/mol Schmelzpunkt: 279…284 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861472 CAS-Nummer: 15996-76-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...